Methyl-3-aminobenzo[b]thiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A731886-100G A731886-1G
103840-788EA 43 USD
103840-788 76727-980 76845-012
Methyl-3-aminobenzo[b]thiophene-2-carboxylate 97%
Methyl 3-aminobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂S
MW: 207.25 g/mol
Boiling Pt: 363.1 °C (760 mm Hg)
Melting Pt: 107…108 °C (760 mm Hg)
Flash Pt: 173.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00206744
CAS Number: 35212-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR