Methyl-3-amino-6-bromopyrazine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-6-bromopyrazine-2-carboxylate

A167117-100G A167117-10G A167117-1G A167117-25G A167117-5G
103806-128EA 6 USD
103806-128 103844-390 76784-092 76784-094 77320-674 77320-676 77320-678
Methyl-3-amino-6-bromopyrazine-2-carboxylate 97%
Methyl-3-amino-6-bromopyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrN₃O₂
MW: 232.03 g/mol
Boiling Pt: 322.4 °C (760 mm Hg)
Melting Pt: 172…177 °C
Flash Pt: 148.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00834964
CAS Number: 6966-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR