Methyl 3-amino-5-methylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-5-methylbenzoate, 3-Amino-5-methyl-benzoic acid methyl ester

A112891-100MG A112891-10G A112891-250MG A112891-25G A112891-5G
103798-128EA 20 USD
103798-128 76718-176 76718-178 76718-180 76718-182 76718-184
Methyl 3-amino-5-methylbenzoate 97%
Methyl 3-amino-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 303.1 °C (760 mm Hg)
Flash Pt: 158.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09263222
CAS Number: 18595-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR