3-Amino-5-cyano-benzoic acid methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-5-cyanobenzoate

A128304-10G A128304-250MG A128304-1G A128304-25G A128304-5G
103801-104EA 24 USD
103801-104 77271-876 77271-878 77298-282 77298-284 77298-286
3-Amino-5-cyano-benzoic acid methyl ester 95%
3-Amino-5-cyano-benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
Boiling Pt: 371.488 °C (760 mm Hg)
Melting Pt: 128 °C
Flash Pt: 178.47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08060133
CAS Number: 199536-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR