Methyl-3-amino-5-bromothiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-5-bromothiophene-2-carboxylate

A125167-100MG A125167-250MG A125167-25G A125167-5G A125167-1G
103800-638EA 37 USD
103800-638 76810-814 76810-816 76810-818 76810-820 77312-782
Methyl-3-amino-5-bromothiophene-2-carboxylate 97%
Methyl-3-amino-5-bromothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
MW: 236.09 g/mol
Boiling Pt: 345.5 °C (760 mm Hg)
Flash Pt: 162.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12031265
CAS Number: 107818-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR