Methyl-3-amino-5-bromobenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-amino-5-bromobenzoate
A185216-1G
A185216-5G
A185216-250MG
A185216-10G
103815-832EA
7
USD
103815-832
103846-400
77084-036
77084-038
77274-254
77567-698
Methyl-3-amino-5-bromobenzoate 97%
Methyl-3-amino-5-bromobenzoate
Precaución: For research use only.
Formula:
C₈H₈BrNO₂ MW: 230.0586 g/mol Boiling Pt: 332 °C (760 mm Hg) Melting Pt: 83…88 °C Flash Pt: 154.775 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07786958 CAS Number: 706791-83-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...