Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate

A108171-100G A108171-10G A108171-250MG A108171-25G A108171-50MG A108171-5G A108171-1G
103797-308EA 10 USD
103797-308 103823-086 76717-936 76717-938 76717-940 76717-942 76717-944 77283-696 77309-930
Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate 97%
Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅Cl₂N₃O₂
MW: 222.03 g/mol
Boiling Pt: 357.3 °C (760 mm Hg)
Melting Pt: 227…230 °C
Flash Pt: 169.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00010431
CAS Number: 1458-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR