Methyl-3-amino-4-methylthiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-4-methyl-2-thiophenecarboxylate, 2-Thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester, Methyl 3-amino-4-methylthiophene-2-carboxylate

A310762-25G A310762-5G A310762-100G A310762-10G A310762-500G
103835-820EA 5 USD
103835-820 76838-594 76838-596 77194-616 77194-618 77194-620
Methyl-3-amino-4-methylthiophene-2-carboxylate 97%
Methyl-3-amino-4-methylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂S
MW: 171.21 g/mol
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00051822
CAS Number: 85006-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR