Methyl-3-amino-4-methylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-4-methylbenzoate

A109946-100G A109946-10G A109946-25G A109946-500G A109946-1000G
103808-030EA 197 USD
103808-030 103823-398 76687-476 76687-478 76687-480 76687-482 77283-910
Methyl-3-amino-4-methylbenzoate 97%
Methyl 3-amino-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 296.3 °C (760 mm Hg)
Melting Pt: 113…117 °C
Flash Pt: 152.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00025206
CAS Number: 18595-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR