Methyl-3-amino-4-methoxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-4-methoxybenzoate, 3-Amino-4-methoxybenzoic acid methyl ester

A178215-100G A178215-10G A178215-1G A178215-5G
103807-304EA 5 USD
103807-304 103845-644 76690-464 76690-466 77083-044 77083-046
Methyl-3-amino-4-methoxybenzoate 98%
Methyl-3-amino-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.19
Boiling Pt: 325.9 °C (760 mm Hg)
Melting Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00017196
CAS Number: 24812-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR