Methyl-3-amino-4-hydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-4-hydroxybenzoate

A266405-100G A266405-10G A266405-250MG A266405-1G A266405-25G A266405-5G
103812-802EA 13 USD
103812-802 103852-704 76722-954 76722-956 76722-958 77092-408 77092-410 77092-412
Methyl-3-amino-4-hydroxybenzoate 98%
Methyl-3-amino-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
Boiling Pt: 326.8 °C (760 mm Hg)
Melting Pt: 141…143 °C
Flash Pt: 151.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659667
CAS Number: 536-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR