Methyl-3-amino-4-fluorobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-amino-4-fluorobenzoate
A238386-100G
A238386-10G
A238386-250MG
A238386-1G
A238386-25G
A238386-5G
103811-228EA
11
USD
103811-228
103851-008
76722-374
76722-376
76722-378
77090-124
77090-126
77090-128
Methyl-3-amino-4-fluorobenzoate 98%
Methyl 3-amino-4-fluorobenzoate
Precaución: for Research Use Only
Formula:
C₈H₈FNO₂ Boiling Pt: 271.1 °C (760 mm Hg) Melting Pt: 66…69 °C Flash Pt: 117.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08689743 CAS Number: 369-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...