Methyl-3-amino-4-bromobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-4-bromobenzoate

A230105-100G A230105-10G A230105-1G A230105-25G
103819-518EA 9 USD
103819-518 103850-418 76722-222 76722-224 77089-270 77089-272
Methyl-3-amino-4-bromobenzoate 97%
Methyl 3-amino-4-bromobenzoate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00553090
CAS Number: 46064-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR