Methyl 3-amino-4-bromo-2-nitrobenzoate 97%

Supplier: Ambeed
Warning

A290733-100MG A290733-250MG A290733-1G
103813-872EA 119 USD
103813-872 103835-014 77239-052 77239-054 77305-582
Methyl 3-amino-4-bromo-2-nitrobenzoate 97%
Methyl 3-amino-4-bromo-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD13677167
CAS Number: 1207175-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR