Methyl-3-amino-2-chloroisonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-2-chloroisonicotinate

A506193-1G A506193-25G A506193-5G A506193-10G A506193-250MG
103828-326EA 11 USD
103828-326 103839-428 76843-404 76843-406 76843-408 77017-246 77017-248
Methyl-3-amino-2-chloroisonicotinate 98%
Methyl-3-amino-2-chloroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.6000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411723
CAS Number: 173435-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR