Methyl 3-amino-2-chlorobenzoate 97%

Supplier: Ambeed
Warning

A101179-250MG A101179-1G A101179-5G
103795-836EA 30 USD
103795-836 103821-522 77008-318 77297-006 77297-008
Methyl 3-amino-2-chlorobenzoate 97%
Methyl 3-amino-2-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Boiling Pt: 298.5±20.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD16036890
CAS Number: 120100-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR