Methyl 3-amino-2-bromobenzoate 97%
Supplier: Ambeed
|
Warning
|
A162823-100MG
A162823-10G
A162823-250MG
A162823-1G
A162823-25G
A162823-5G
103805-660EA
26
USD
103805-660
103843-914
77273-398
77273-400
77273-402
77319-988
77319-990
77319-992
Methyl 3-amino-2-bromobenzoate 97%
Methyl 3-amino-2-bromobenzoate
Precaución: for Research Use Only
Formula:
C₈H₈BrNO₂ Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03844437 CAS Number: 106896-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...