Methyl-3-amino-2,2-dimethylpropanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-2,2-dimethylpropanoate hydrochloride

A129389-1G A129389-5G A129389-10G A129389-250MG
103801-274EA 44 USD
103801-274 77313-656 77313-658 77881-031 77881-032
Methyl-3-amino-2,2-dimethylpropanoate hydrochloride 97%
Methyl-3-amino-2,2-dimethylpropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₂
Melting Pt: 167…168 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10758092
CAS Number: 177269-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR