Methyl 3-amino-1H-indazole-6-carboxylate 96%

Supplier: Ambeed
Warning

A101289-100MG A101289-250MG
103795-894EA 368 USD
103795-894 77233-072 77233-074
Methyl 3-amino-1H-indazole-6-carboxylate 96%
Methyl 3-amino-1H-indazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₉N₃O₂
MW: 191.19 g/mol
Storage Temperature: Refrigerator
CAS Number: 1279865-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR