Methyl-3-(acetylamino)-2-thiophenecarboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-acetamidothiophene-2-carboxylate
A303463-100MG
A303463-1G
A303463-5G
103814-374EA
16
USD
103814-374
103835-542
76723-644
76838-228
76838-230
Methyl-3-(acetylamino)-2-thiophenecarboxylate 97%
Methyl-3-(acetylamino)-2-thiophenecarboxylate
Precaución: for Research Use Only
Formula:
C₈H₉NO₃S MW: 199.23 g/mol Boiling Pt: 399.9 °C (760 mm Hg) Melting Pt: 94…95 °C Flash Pt: 195.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100044 CAS Number: 22288-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...