Methyl 3,6-dichloropyridazine-4-carboxylate 97%

Supplier: Ambeed
Warning

A166657-10G A166657-250MG A166657-25G A166657-1G A166657-5G
103806-086EA 18 USD
103806-086 103844-350 76824-334 76824-336 77300-844 77320-624 77320-626
Methyl 3,6-dichloropyridazine-4-carboxylate 97%
Methyl 3,6-dichloropyridazine-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O₂
MW: 207.01 g/mol
Boiling Pt: 357.3 °C (760 mm Hg)
Flash Pt: 169.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12546173
CAS Number: 286946-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR