Methyl-3,5-dimethylbenzoate 97%
Supplier: Ambeed
|
Warning
|
A265415-100G
A265415-10G
A265415-250MG
A265415-500G
A265415-1G
A265415-25G
103812-730EA
7
USD
103812-730
103852-624
76722-916
76722-918
76722-920
76722-922
77092-312
77092-314
Methyl-3,5-dimethylbenzoate 97%
Methyl 3,5-dimethylbenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₂ Boiling Pt: 242.9 °C (760 mm Hg) Melting Pt: 31…33 °C Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077971 CAS Number: 25081-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...