Methyl-3,5-dihydroxybenzoate 98%

Supplier: Ambeed
Warning

A264747-1000G A264747-100G A264747-10G A264747-500G A264747-25G A264747-5G
103852-580EA 5 USD
103852-580 77250-224 77250-226 77250-228 77250-230 77304-900 77304-902
Methyl-3,5-dihydroxybenzoate 98%
Methyl-3,5-dihydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₈O₄
Boiling Pt: 349.4 °C (760 mm Hg)
Melting Pt: 164…167 °C
Flash Pt: 147.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00002289
CAS Number: 2150-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR