Methyl 3,5-dichloropyrazine-2-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A157961-100G
A157961-100MG
A157961-10G
A157961-250MG
A157961-1G
A157961-25G
A157961-5G
103805-070EA
19
USD
103805-070
103843-276
77028-474
77028-476
77028-478
77028-480
77319-114
77319-116
77319-118
Methyl 3,5-dichloropyrazine-2-carboxylate 98%
Methyl 3,5-dichloropyrazine-2-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₄Cl₂N₂O₂ MW: 207.01 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09953610 CAS Number: 330786-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...