Methyl 3,5-dichloro-4-hydroxybenzoate 97%

Supplier: Ambeed
Warning

A141364-100G A141364-10G A141364-250MG A141364-1G A141364-25G A141364-5G
103802-982EA 6 USD
103802-982 77109-816 77109-818 77109-820 77299-208 77316-134 77316-136
Methyl 3,5-dichloro-4-hydroxybenzoate 97%
Methyl 3,5-dichloro-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₃
MW: 221.03 g/mol
Boiling Pt: 302.7 °C (760 mm Hg)
Melting Pt: 122…125 °C
Flash Pt: 136.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00016419
CAS Number: 3337-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR