Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3,4-dihydro-2H-benzo[b][1,4]oxazine-7-carboxylate
A362116-100MG
A362116-10G
A362116-1G
A362116-250MG
103826-300EA
69
USD
103826-300
103837-314
77278-330
77278-332
77306-728
77748-720
Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate 97%
Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁NO₃ MW: 193.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11848180 CAS Number: 142166-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...