Methyl 3,4-dihydro-2H-benzo[b][1,4]oxazine-5-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A175189-250MG
A175189-1G
103807-006EA
317
USD
103807-006
103845-312
76872-554
77301-366
Methyl 3,4-dihydro-2H-benzo[b][1,4]oxazine-5-carboxylate 97%
Methyl 3,4-dihydro-2H-benzo[b][1,4]oxazine-5-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁NO₃ MW: 193.2 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11878403 CAS Number: 121591-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...