Methyl-3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate

A103531-100MG A103531-250MG A103531-5G A103531-1G
103796-568EA 78 USD
103796-568 103822-274 77270-358 77270-360 77283-114 77309-148
Methyl-3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate 97%
Methyl-3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
MW: 193.2000 g/mol
Flash Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD08544342
CAS Number: 758684-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR