Methyl-3,4,5-trifluorobenzenecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3,4,5-trifluorobenzoate

A347007-1G A347007-25G A347007-100G A347007-10G A347007-500G A347007-5G
103825-948EA 9 USD
103825-948 103836-938 76840-006 76840-008 77003-216 77003-218 77003-220 77306-578
Methyl-3,4,5-trifluorobenzenecarboxylate 98%
Methyl-3,4,5-trifluorobenzenecarboxylate

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 211.1 °C (760 mm Hg)
Flash Pt: 83.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06200977
CAS Number: 773873-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR