Methyl-3,3-dimethoxypropionate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3,3-dimethoxypropanoate, Methyl 3,3-dimethoxypropionate
A127066-100G
A127066-10G
A127066-500G
A127066-5G
A127066-25G
76782-086EA
50.93
USD
76782-086
76782-088
76782-090
77298-170
77313-156
Methyl-3,3-dimethoxypropionate 97%
Methyl 3,3-dimethoxypropionate
Precaución: for Research Use Only
Formula:
C₆H₁₂O₄ MW: 148.16 g/mol Boiling Pt: 178.8 °C (760 mm Hg) Flash Pt: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010650 CAS Number: 7424-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...