Methyl trifluoropyruvate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Methyl 3,3,3-trifluoropyruvate, Methyl 3,3,3-trifluoro-2-oxopropanoate
A383524-5G
A383524-1G
A383524-25G
103826-742EA
9
USD
103826-742
103837-778
76841-110
77306-940
77306-942
Methyl trifluoropyruvate 95%
Methyl trifluoropyruvate
Precaución: for Research Use Only
Formula:
C₄H₃F₃O₃ MW: 156.06 g/mol Boiling Pt: 85 °C (760 mm Hg) Flash Pt: 26 °C Storage Temperature: Freezer |
MDL Number:
MFCD00114936 CAS Number: 13089-11-7 UN: 1993 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...