Methyl-3-(trifluoromethyl)picolinate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-(trifluoromethyl)picolinate

A426989-1G A426989-25G A426989-5G A426989-10G A426989-250MG
103827-480EA 23 USD
103827-480 103838-532 76842-188 76842-190 76842-192 76979-288 76979-290
Methyl-3-(trifluoromethyl)picolinate 95%
Methyl-3-(trifluoromethyl)picolinate

Precaución: For research use only

Formula: C₈H₆F₃NO₂
MW: 205.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698094
CAS Number: 588702-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR