Methyl 4-Methylcinnamate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methylcinnamic acid methyl ester, Methyl 3-(p-tolyl)acrylate

A282610-25G A282610-10G A282610-250MG A282610-1G A282610-100G
103813-594EA 9 USD
103813-594 103853-546 76837-244 77099-184 77099-186 77305-436 77685-801
Methyl 4-Methylcinnamate 97%
Methyl 4-Methylcinnamate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 272.9 °C (760 mm Hg)
Flash Pt: 152.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00460746
CAS Number: 7560-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR