Methyl 3-(piperidin-4-yl)benzoate hydrochloride 97%

Supplier: Ambeed
Warning

A117672-100MG A117672-250MG A117672-1G
103798-924EA 109 USD
103798-924 103824-812 76983-062 76983-064 77297-554
Methyl 3-(piperidin-4-yl)benzoate hydrochloride 97%
Methyl 3-(piperidin-4-yl)benzoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₈ClNO₂
MW: 255.75 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02179142
CAS Number: 726185-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR