Methyl 3-(methylamino)propanoate 95%

Supplier: Ambeed
Warning

A218755-250MG A218755-1G A218755-5G
103818-744EA 56 USD
103818-744 103849-614 76826-100 77303-510 77303-512
Methyl 3-(methylamino)propanoate 95%
Methyl 3-(methylamino)propanoate

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 50 °C (11 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD11505527
CAS Number: 24549-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR