Methyl 3-(bromomethyl)-4-methoxybenzoate 98%

Supplier: Ambeed
Danger

A317866-1G A317866-250MG
103825-152EA 276 USD
103825-152 103836-082 76838-928 76989-074
Methyl 3-(bromomethyl)-4-methoxybenzoate 98%
Methyl 3-(bromomethyl)-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
MW: 259.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14706047
CAS Number: 142031-88-7
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR