Methyl-3-(aminomethyl)benzoate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-(aminomethyl)benzoate hydrochloride

A187926-10G A187926-1G
103816-054EA 5 USD
103816-054 103846-646 76721-070 77084-364
Methyl-3-(aminomethyl)benzoate hydrochloride 95%
Methyl-3-(aminomethyl)benzoate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₂
MW: 201.65 g/mol
Boiling Pt: 279.8 °C (760 mm Hg)
Melting Pt: 179…181 °C
Flash Pt: 139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039173
CAS Number: 17841-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR