Methyl-3-(4-hydroxyphenyl)acrylate 98%
Supplier: Ambeed
|
Warning
|
A225252-100G
A225252-10G
A225252-500G
A225252-25G
A225252-5G
103819-168EA
6
USD
103819-168
103850-062
76722-094
76722-096
76722-098
77088-746
77088-748
Methyl-3-(4-hydroxyphenyl)acrylate 98%
Methyl-3-(4-hydroxyphenyl)acrylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₀O₃ Melting Pt: 144…145 °C Flash Pt: 132.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00157167 CAS Number: 3943-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...