Methyl-3-(4-(bromomethyl)phenyl)acrylate 97%
Supplier: Ambeed
|
Danger
|
A111036-100G
A111036-10G
A111036-25G
A111036-1G
A111036-5G
103797-782EA
9
USD
103797-782
103823-590
76869-714
76869-716
76869-718
77284-080
77284-082
Methyl-3-(4-(bromomethyl)phenyl)acrylate 97%
Methyl-3-(4-(bromomethyl)phenyl)acrylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₁BrO₂ Boiling Pt: 333.9 °C (760 mm Hg) Melting Pt: 58…61 °C Flash Pt: 155.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02684186 CAS Number: 946-99-6 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...