Methyl 3-(3-bromophenyl)propanoate 95%
Supplier: Ambeed
|
Warning
|
A211942-10G
A211942-1G
A211942-25G
A211942-5G
103818-148EA
13
USD
103818-148
103848-844
76873-694
77087-276
77087-278
77087-280
Methyl 3-(3-bromophenyl)propanoate 95%
Methyl 3-(3-bromophenyl)propanoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁BrO₂ MW: 243.09 g/mol Boiling Pt: 287.3±15.0 °C (760 mm Hg) Flash Pt: 127.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD11855856 CAS Number: 151583-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...