Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid methyll ester, Methyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate

A131100-100G A131100-500G
103808-416EA 121 USD
103808-416 76811-194 77549-560
Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate 97%
Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate

Precaución: for Research Use Only

Formula: C₁₈H₂₈O₃
MW: 292.42 g/mol
Melting Pt: 60…67 °C
Flash Pt: 124.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00210461
CAS Number: 6386-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR