Methyl 3-(2,6-difluorophenylsulfonamido)-2-fluorobenzoate 97%

Supplier: Ambeed
Warning

A248791-100MG A248791-250MG A248791-1G A248791-5G
103811-838EA 52 USD
103811-838 103851-672 77165-932 77165-934 77304-460 77304-462
Methyl 3-(2,6-difluorophenylsulfonamido)-2-fluorobenzoate 97%
Methyl 3-(2,6-difluorophenylsulfonamido)-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₃NO₄S
Storage Temperature: Ambient
MDL Number: MFCD18447698
CAS Number: 1195768-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR