Methyl 3-(1H-pyrrol-2-yl)propanoate 98%

Supplier: Ambeed
Warning

A152832-100MG A152832-250MG A152832-1G A152832-25G A152832-5G
103804-438EA 23 USD
103804-438 103842-610 76871-682 76871-684 77299-984 77318-244 77318-246
Methyl 3-(1H-pyrrol-2-yl)propanoate 98%
Methyl 3-(1H-pyrrol-2-yl)propanoate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MW: 153.178 g/mol
Boiling Pt: 250.6 °C (760 mm Hg)
Flash Pt: 105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09841003
CAS Number: 69917-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR