Methyl-2-pyrazinecarboxylate 97%

Supplier: Ambeed
Warning

A203314-1G A203314-25G A203314-5G A203314-100G A203314-10G
103817-456EA 8 USD
103817-456 103848-110 77086-242 77086-244 77086-246 77112-252 77112-254
Methyl-2-pyrazinecarboxylate 97%
Methyl-2-pyrazinecarboxylate

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
Boiling Pt: 220.4 °C (760 mm Hg)
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00014611
CAS Number: 6164-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR