Methyl 2-pyrazineacetate 98%

Supplier: Ambeed
Warning

A166888-10G A166888-250MG A166888-25G A166888-5G
103806-110EA 25 USD
103806-110 103844-372 77191-304 77191-306 77300-856 77320-656
Methyl 2-pyrazineacetate 98%
Methyl 2-pyrazineacetate

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₇H₈N₂O₂
Boiling Pt: 226.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04038671
CAS Number: 370562-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR