Methyl-2-phenylacrylate ≥95% stabilized

Supplier: Ambeed
Warning

Synonyms: Methyl 2-phenylacrylate

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A148581-100MG A148581-1G A148581-250MG
103803-944EA 166 USD
103803-944 103842-060 77272-870 77299-672 77567-086
Methyl-2-phenylacrylate ≥95% stabilized
Methyl-2-phenylacrylate

Precaución: For research use only.

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 256 °C (760 mm Hg)
Flash Pt: 145.4 °C
Storage Temperature: Freezer
MDL Number: MFCD09032492
CAS Number: 1865-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR