Methyl 2-oxopiperidine-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-oxoisonipecotate, 4-(Methoxycarbonyl)-2-oxopiperidine

A106805-10G A106805-250MG A106805-25G A106805-1G A106805-5G
103797-078EA 26 USD
103797-078 103822-836 76970-468 76970-470 76970-472 77309-686 77309-688
Methyl 2-oxopiperidine-4-carboxylate 95%
Methyl 2-oxopiperidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃
MW: 157.167 g/mol
Boiling Pt: 316.0±35.0 °C (760 mm Hg)
Flash Pt: 144.9 °C
Storage Temperature: Ambient
MDL Number: MFCD17012745
CAS Number: 25504-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR