Methyl oxindole-6-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-Oxoindoline-6-carboxylate, 2-Oxoindoline-6-carboxylic acid methyll ester

A234849-1G A234849-100G A234849-10G A234849-250MG A234849-500G A234849-25G A234849-5G
103810-980EA 8 USD
103810-980 103850-738 77089-738 77275-828 77275-830 77275-832 77275-834 77303-998 77304-000
Methyl oxindole-6-carboxylate 98%
Methyl oxindole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MW: 191.1800 g/mol
Boiling Pt: 388.1 °C (760 mm Hg)
Melting Pt: 184…190 °C
Flash Pt: 188.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03095196
CAS Number: 14192-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR