Methyl-2-oxocyclohexanecarboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-oxocyclohexanecarboxylate
A153451-25G
A153451-1G
A153451-5G
A153451-10G
103804-518EA
8
USD
103804-518
103842-690
77259-344
77318-350
77318-352
77567-228
Methyl-2-oxocyclohexanecarboxylate 95%
Methyl-2-oxocyclohexanecarboxylate
Precaución: For research use only.
Formula:
C₈H₁₂O₃ MW: 156.1791 g/mol Boiling Pt: 230.8 °C (760 mm Hg) Flash Pt: 94.9 °C Storage Temperature: Freezer |
MDL Number:
MFCD00077699 CAS Number: 41302-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...