Methyl phenylglyoxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl benzoylformate, Methyl 2-oxo-2-phenylacetate

A139827-100G A139827-10G A139827-500G A139827-25G
103808-596EA 97 USD
103808-596 76996-826 76996-828 76996-830 77315-870
Methyl phenylglyoxylate 97%
Methyl phenylglyoxylate

Precaución: for Research Use Only

Formula: C₉H₈O₃
MW: 164.1600 g/mol
Boiling Pt: 247 °C (760 mm Hg)
Melting Pt: 16 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008443
CAS Number: 15206-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR